N,N-Diethylacetoacetamide ≥97%
Dodavatel: Thermo Fisher Scientific
Purity: 97%
CAS No.: 2235-46-3
Molecular Formula: CH3COCH2CON(CH2CH3)2
Formula Weight: 157.20
Flash Point: 94°(201°F)
Refractive Index: 1.4720
Melting Point:
Vzorec:
C₈H₁₅NO₂ Bod varu: 213…218 °C Bod tání: < –73 °C Hustota: 0,990 Bod vzplanutí: 94 °C (201 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00026728 CAS číslo: 2235-46-3 EINECS: 218-792-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...