Tisk…

Dipropyleneglycol dimethyl ether (mixture of isomers), Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Dipropylene glycol dimethyl ether

M-Clarity™ quality level = MQ100

520462-1L 520462-5L
SIAL520462-1LEA 4530 CZK
SIAL520462-1L SIAL520462-5L
Dipropyleneglycol dimethyl ether (mixture of isomers), Sigma-Aldrich®
Dipropyleneglycol dimethyl ether (mixture of isomers)
Vzorec: C₈H₁₈O₃
MDL Number: MFCD00210047
CAS číslo: 111109-77-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR