Tisk…

Dipropyleneglycol (mixture of isomers), Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Dipropylene glycol

M-Clarity™ quality level = MQ200

D215554-25G D215554-3KG D215554-500G
SIALD215554-25GEA 1220 CZK
SIALD215554-25G SIALD215554-3KG SIALD215554-500G
Dipropyleneglycol (mixture of isomers), Sigma-Aldrich®
Dipropyleneglycol (mixture of isomers)
Vzorec: C₆H₁₄O₃
MW: 134,18 g/mol
MDL Number: MFCD00051023
CAS číslo: 25265-71-8
EINECS: 246-770-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR