Tisk…

Ethyl (ethoxymethylene)cyanoacetate 98%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: Ethyl 2-Cyano-3-Ethoxyacrylate

118481000 118485000 118480010
ACRO118481000EA 1490 CZK
ACRO118481000 ACRO118485000 ACRO118480010
Ethyl (ethoxymethylene)cyanoacetate 98%
Ethyl (ethoxymethylene)cyanoacetate
Vzorec: C₈H₁₁NO₃
MW: 169,18 g/mol
Bod varu: 190…191 °C (30 mmHg)
Bod tání: 48…52 °C
Bod vzplanutí: 130 °C
Storage Temperature: Ambient
MDL Number: MFCD00009136
CAS číslo: 94-05-3
EINECS: 202-299-5
ADR: 6.1,III

Objednat

Výsledky specifikačního testu

Appearance White to yellow to yellow-brown Powder or crystals or flakes or crystalline solid
Infrared spectrum Conforms
Melting point 47 to 53 °C
GC ≥99.5 %
Color scale ≤10 APHA
Residue after evaporation ≤0.001 %
Substance darkened by H2SO4 passes test
Sulfur compounds (as S) ≤0.003 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR