4-Amino-3-hydrazino-5-mercapto-1,2,4-triazole, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
162892-100G
162892-5G
SIAL162892-100GEA
27410
CZK
SIAL162892-100G
SIAL162892-5G
4-Amino-3-hydrazino-5-mercapto-1,2,4-triazole, Sigma-Aldrich®
4-Amino-3-hydrazino-5-mercapto-1,2,4-triazole
Vzorec:
C₂H₆N₆S |
MDL Number:
MFCD00003098 CAS číslo: 1750-12-5 EINECS: 217-135-8 UN: 1325 ADR: 4.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...