Tisk…

3-Methyl-3-oxetanemethanol, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 3-Methyl-3-oxethanemethanol

M-Clarity™ quality level = MQ200

277681-10G 277681-50G
SIAL277681-10GEA 1520 CZK
SIAL277681-10G SIAL277681-50G
3-Methyl-3-oxetanemethanol, Sigma-Aldrich®
3-Methyl-3-oxetanemethanol
Vzorec: C₅H₁₀O₂
MW: 102,13 g/mol
MDL Number: MFCD00010273
CAS číslo: 3143-02-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR