Tisk…

(±)-2-Pyrrolidinone-5-carboxylic acid ≥99%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: 5-Oxopyrrolidine-2-carboxylic acid, DL-2-Pyrrolidone-5-carboxylic acid, DL-Pyroglutamic acid

L09424.18 L09424.30
L09424.18EA 913 CZK
L09424.18 L09424.30
(±)-2-Pyrrolidinone-5-carboxylic acid ≥99%
(±)-2-Pyrrolidinone-5-carboxylic acid

Purity: 99% CAS No.: 149-87-1 Molecular Formula: C5H7NO3 Formula Weight: 129.12 Melting Point: 183-185° Beilstein Reference: 82131

Vzorec: C₅H₇NO₃
MW: 129.12 g/mol
Bod tání: 183…185 °C
Storage Temperature: Ambient
MDL Number: MFCD00064322
CAS číslo: 149-87-1
EINECS: 205-748-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR