Pyrrole-3-carboxylic acid ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
H55383.MD
H55383.03
H55383.06
H55383.MDEA
1450
CZK
H55383.MD
H55383.03
H55383.06
Pyrrole-3-carboxylic acid ≥98%
Pyrrole-3-carboxylic acid
Purity: 98+%
CAS No.: 931-03-3
Molecular Formula: C5H5NO2
Formula Weight: 111.10
Beilstein Reference: 108642
MDL No.: MFCD00800594
Vzorec:
C₅H₅NO₂ Storage Temperature: Ambient |
MDL Number:
MFCD00800594 CAS číslo: 931-03-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...