Tisk…

Pyrrole-2-carboxylic acid ≥99%

Dodavatel: Thermo Scientific
Varování

L13756.03 L13756.06 L13756.14
L13756.03EA 1470 CZK
L13756.03 L13756.06 L13756.14
Pyrrole-2-carboxylic acid ≥99%
Pyrrole-2-carboxylic acid

Purity: 99% CAS No.: 634-97-9 Molecular Formula: C5H5NO2 Formula Weight: 111.10 Melting Point: ca 205° subl. Beilstein Reference: 80825

Vzorec: C₅H₅NO₂
MW: 111,10 g/mol
Bod tání: ∼ 205 °C (subl.)
Storage Temperature: Ambient
MDL Number: MFCD00005219
CAS číslo: 634-97-9
EINECS: 211-221-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR