Tisk…

5-Fluoroindoline ≥97%

Dodavatel: Thermo Scientific
Varování

H54158.MD H54158.03 H54158.06
H54158.MDEA 1360 CZK
H54158.MD H54158.03 H54158.06
5-Fluoroindoline ≥97%
5-Fluoroindoline

Purity: 97% CAS No.: 2343-22-8 Molecular Formula: C8H8FN Formula Weight: 137.16 Storage Temperature: 2°C to 8°C

Vzorec: C₈H₈FN
MW: 137,16 g/mol
MDL Number: MFCD00214461
CAS číslo: 2343-22-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR