Tisk…

5-Bromoindoline, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

642371-1G 642371-5G
SIAL642371-1GEA 0 CZK
SIAL642371-1G SIAL642371-5G
5-Bromoindoline, Sigma-Aldrich®
5-Bromoindoline
Vzorec: C₈H₈BrN
MW: 198,06 g/mol
MDL Number: MFCD00027410
CAS číslo: 22190-33-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR