Tisk…

3-Nitro-L-tyrosine, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

N7389-10G N7389-25G N7389-5G
SIALN7389-10GEA 7130 CZK
SIALN7389-10G SIALN7389-25G SIALN7389-5G
3-Nitro-L-tyrosine, Sigma-Aldrich®
3-Nitro-L-tyrosine
Vzorec: C₉H₁₀N₂O₅
MDL Number: MFCD00007123
CAS číslo: 621-44-3
EINECS: 210-688-6

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR