Tisk…

Triethyl-3-phosphonopropionate ≥98%

Dodavatel: Thermo Fisher Scientific

L10190.09 L10190.18 L10190.TW
L10190.09EA 2150 CZK
L10190.09 L10190.18 L10190.TW
Triethyl-3-phosphonopropionate ≥98%
Triethyl-3-phosphonopropionate

Purity: 98% CAS No.: 3699-67-0 Molecular Formula: C9H19O5P Formula Weight: 238.22 Flash Point: 110°(230°F) Refractive Index: 1.4330 Boiling Point: 127-129°/2mm Beilstein Reference: 1790966 MDL No.: MFCD00015167

Vzorec: C₉H₁₉O₅P
Bod varu: 127…129 °C (2 mmHg)
Hustota: 1,094
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00015167
CAS číslo: 3699-67-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR