Triethyl phosphonoacetate ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
Ethyl 2-(diethoxyphosphoryl)acetate, Diethyl [(ethoxycarbonyl)methyl]phosphonate
A14120.0E
A14120.22
A14120.36
A14120.0EEA
22450
CZK
A14120.0E
A14120.22
A14120.36
Triethyl phosphonoacetate ≥98%
Triethyl phosphonoacetate
Purity: 98+%
CAS No.: 867-13-0
Molecular Formula: C8H17O5P
Formula Weight: 224.19
Flash Point: 165°(329°F)
Refractive Index: 1.4310
Melting Point: -24°
Boiling Point: 260-262°
Beilstein Reference: 1343714
MDL No.: MFCD00009177
Vzorec:
C₈H₁₇O₅P MW: 224,19 g/mol Bod varu: 260…262 °C Bod tání: –24 °C Hustota: 1,125 Bod vzplanutí: 165 °C (329 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009177 CAS číslo: 867-13-0 EINECS: 212-757-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...