Octylphosphonic acid ≥99%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
OPA, 1-Octylphosphonic acid, n-Octylphosphonic acid
L18586.06
L18586.03
L18586.06EA
8620
CZK
L18586.06
L18586.03
Octylphosphonic acid ≥99%
Octylphosphonic acid
Purity: 99%
CAS No.: 4724-48-5
Molecular Formula: C8H19O3P
Formula Weight: 194.21
Melting Point: 100-102°
Beilstein Reference: 1764242
Vzorec:
C₈H₁₉O₃P MW: 194,21 g/mol Bod tání: 100…102 °C Storage Temperature: Ambient |
MDL Number:
MFCD00015841 CAS číslo: 4724-48-5 EINECS: 225-218-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...