Octylphosphonic acid ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
20651.03
20651.06
20651.03EA
2200
CZK
20651.03
20651.06
Octylphosphonic acid ≥98%
Octylphosphonic acid
Purity: 98%
CAS No.: 4724-48-5
Molecular Formula: C8H19O3P
Formula Weight: 194.21
Physical Form: Crystalline
Melting Point: 100-102°
Beilstein Reference: 1764242
MDL No.: MFCD00015841
Vzorec:
C₈H₁₉O₃P Bod tání: 100…102 °C Storage Temperature: Ambient |
MDL Number:
MFCD00015841 CAS číslo: 4724-48-5 EINECS: 225-218-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...