Octadecylphosphonic acid ≥97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
20645.03
20645.06
20645.03EA
4330
CZK
20645.03
20645.06
Octadecylphosphonic acid ≥97%
Octadecylphosphonic acid
Purity: 97%
CAS No.: 4724-47-4
Molecular Formula: C18H39O3P
Formula Weight: 334.48
Physical Form: Crystalline
Appearance: White to off-white
Melting Point: 92-98°
MDL No.: MFCD00015838
Vzorec:
C₁₈H₃₉O₃P Bod tání: 92…98 °C Storage Temperature: Ambient |
MDL Number:
MFCD00015838 CAS číslo: 4724-47-4 EINECS: 225-216-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...