Tisk…

Decylphosphonic acid ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

L12019.03 L12019.06
L12019.03EA 1700 CZK
L12019.03 L12019.06
Decylphosphonic acid ≥98%
Decylphosphonic acid

Purity: 98% CAS No.: 6874-60-8 Molecular Formula: CH3(CH2)9PO3H2 Formula Weight: 222.27 Melting Point: 100-101° Beilstein Reference: 1771572 UN No.: 3261 MDL No.: MFCD00015830

Vzorec: C₁₀H₂₃O₃P
Bod tání: 100…101 °C
Storage Temperature: Ambient
MDL Number: MFCD00015830
CAS číslo: 6874-60-8
EINECS: 229-975-2
UN: 3261
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR