Butylphosphonic acid ≥98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
1-Butylphosphonic acid, Butylphosphonic Acid
A17983.06
A17983.14
A17983.06EA
4740
CZK
A17983.06
A17983.14
Butylphosphonic acid ≥98%
Butylphosphonic acid
Purity: 98%
CAS No.: 3321-64-0
Molecular Formula: C4H11O3P
Formula Weight: 138.10
Odour: Odourless
Melting Point: 105-107°
Beilstein Reference: 1749026
UN No.: 3261
MDL No.: MFCD00012295
Vzorec:
C₄H₁₁O₃P Bod tání: 105…107 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012295 CAS číslo: 3321-64-0 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...