Tisk…

3-Bromobenzo[b]thiophene 95%

Dodavatel: Thermo Fisher Scientific
Varování

H32948.09 H32948.18
H32948.09EA 2710 CZK
H32948.09 H32948.18
3-Bromobenzo[b]thiophene 95%
3-Bromobenzo[b]thiophene

Purity: 95% CAS No.: 7342-82-7 Molecular Formula: C8H5BrS Formula Weight: 213.10 Refractive Index: 1.6680 Boiling Point: 72-74°/0.27mm MDL No.: MFCD00023009

Vzorec: C₈H₅BrS
Bod varu: 72…74 °C (0,27 mmHg)
Hustota: 1,629
Storage Temperature: Ambient
MDL Number: MFCD00023009
CAS číslo: 7342-82-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR