Tisk…

Potassium tetrachloroaurate ≥99.995% (trace metals basis), Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

450235-1G 450235-250MG
SIAL450235-1GEA 10830 CZK
SIAL450235-1G SIAL450235-250MG
Potassium tetrachloroaurate ≥99.995% (trace metals basis), Sigma-Aldrich®
Potassium tetrachloroaurate
Vzorec: AuCl₄K
MW: 377,88 g/mol
MDL Number: MFCD00043088
CAS číslo: 13682-61-6
EINECS: 237-190-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR