Tisk…

2-(4-tert-Butylphenyl)ethanol 96%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: 2-(4-(tert-Butyl)phenyl)ethanol

L13611.06 L13611.14
L13611.06EA 1740 CZK
L13611.06 L13611.14
2-(4-tert-Butylphenyl)ethanol 96%
2-(4-tert-Butylphenyl)ethanol

Purity: 96% CAS No.: 5406-86-0 Molecular Formula: C12H18O Formula Weight: 178.28 Refractive Index: 1.5209 Melting Point: 31-35° Boiling Point: 141-143°/15mm Beilstein Reference: 2246595 UN No.: 3077 MDL No.: MFCD00236023

Vzorec: C₁₂H₁₈O
MW: 178,28 g/mol
Bod varu: 141…143 °C (15 mmHg)
Bod tání: 31…35 °C
Hustota: 0,975
Storage Temperature: Ambient
MDL Number: MFCD00236023
CAS číslo: 5406-86-0
EINECS: 410-020-5
ADR: 9,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR