Tisk…

3-Chloro-4-methoxyphenylboronic acid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 3-Chloro-4-methoxybenzeneboronic acid, (3-Chloro-4-methoxyphenyl)boronic acid

M-Clarity™ quality level = MQ100

564486-1G 564486-5G
SIAL564486-1GEA 1410 CZK
SIAL564486-1G SIAL564486-5G
3-Chloro-4-methoxyphenylboronic acid, Sigma-Aldrich®
3-Chloro-4-methoxyphenylboronic acid
Vzorec: C₇H₈BClO₃
MW: 186,40 g/mol
MDL Number: MFCD04039888
CAS číslo: 175883-60-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR