Tisk…

2-Chloro-6-methoxyphenylboronic acid, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

512753-1G 512753-5G
SIAL512753-1GEA 0 CZK
SIAL512753-1G SIAL512753-5G
2-Chloro-6-methoxyphenylboronic acid, Sigma-Aldrich®
2-Chloro-6-methoxyphenylboronic acid
Vzorec: C₇H₈BClO₃
MW: 186,4000 g/mol
MDL Number: MFCD04039878
CAS číslo: 385370-80-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR