2-Nitrophenyl octyl ether, Supelco®
Dodavatel: Merck
Synonyms:
1-Nitro-2-(n-octyloxy)benzene, 1-Nitro-2-(octyloxy)benzene, 2-(n-Octyloxy)nitrobenzene
M-Clarity™ quality level = MQ100
73732-100ML
73732-25ML
73732-5ML
73732-100MLEA
30310
CZK
73732-100ML
73732-25ML
73732-5ML
2-Nitrophenyl octyl ether, Supelco®
2-Nitrophenyl octyl ether
Vzorec:
C₁₄H₂₁NO₃ MW: 251,32 g/mol |
MDL Number:
MFCD00014693 CAS číslo: 37682-29-4 EINECS: 253-623-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...