Tisk…

2-Nitrophenyl octyl ether ≥98%

Dodavatel: Thermo Fisher Scientific

Synonyms: 1-Nitro-2-(n-octyloxy)benzene, 1-Nitro-2-(octyloxy)benzene, 2-(n-Octyloxy)nitrobenzene

A17320.14 A17320.06
A17320.14EA 7010 CZK
A17320.14 A17320.06
2-Nitrophenyl octyl ether ≥98%
2-Nitrophenyl octyl ether

Purity: 98% CAS No.: 37682-29-4 Molecular Formula: C14H21NO3 Formula Weight: 251.32 Flash Point: 110°(230°F) Refractive Index: 1.5110 Boiling Point: 163-165°/2mm Beilstein Reference: 2529905 MDL No.: MFCD00014693

Vzorec: C₁₄H₂₁NO₃
Bod varu: 163…165 °C (2 mmHg)
Hustota: 1,04
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00014693
CAS číslo: 37682-29-4
EINECS: 253-623-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR