Tisk…

1,4-Dioxane-2,5-dione ≥97%

Dodavatel: Thermo Scientific
Varování

Synonyms: Glycolide, 1,4-Dioxane-2,5-dione

B20096.06 B20096.14
B20096.06EA 1480 CZK
B20096.06 B20096.14
1,4-Dioxane-2,5-dione ≥97%
1,4-Dioxane-2,5-dione

Purity: 97% CAS No.: 502-97-6 Molecular Formula: C4H4O4 Formula Weight: 116.07 Melting Point: 81-83° MDL No.: MFCD00081108

Vzorec: C₄H₄O₄
MW: 116,07 g/mol
Bod tání: 81…83 °C
Storage Temperature: Ambient
MDL Number: MFCD00081108
CAS číslo: 502-97-6
EINECS: 207-954-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR