Tisk…

3-Buten-1-amine 97%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

L20157.03 L20157.06
L20157.03EA 3880 CZK
L20157.03 L20157.06
3-Buten-1-amine 97%
3-Buten-1-amine

Purity: 97% CAS No.: 2524-49-4 Molecular Formula: C4H9N Formula Weight: 71.12 Flash Point: -5°(23°F) Refractive Index: 1.4240 Odour: Amine-like Boiling Point: 74-77° Beilstein Reference: 1735803 UN No.: 2733 MDL No.: MFCD03425859

Vzorec: C₄H₉N
Bod varu: 74…77 °C
Hustota: 0,777
Bod vzplanutí: –5 °C (23 °F)
Storage Temperature: Ambient
MDL Number: MFCD03425859
CAS číslo: 2524-49-4
ADR: 3,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR