Benzyl carbamate, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
B18200-25G
B18200-100G
SIALB18200-25GEA
1450
CZK
SIALB18200-25G
SIALB18200-100G
Benzyl carbamate, Sigma-Aldrich®
Benzyl carbamate
Vzorec:
C₈H₉NO₂ MW: 151,17 g/mol |
MDL Number:
MFCD00007965 CAS číslo: 621-84-1 EINECS: 210-710-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...