Benzyl carbamate ≥99%
Dodavatel: Thermo Scientific
A11569.14
A11569.22
A11569.30
A11569.14EA
1140
CZK
A11569.14
A11569.22
A11569.30
Benzyl carbamate ≥99%
Benzyl carbamate
Purity: 99%
CAS No.: 621-84-1
Molecular Formula: H2NCO2CH2C6H5
Formula Weight: 151.17
Melting Point: 85-89°
Beilstein Reference: 1865635
Vzorec:
C₈H₉NO₂ MW: 151,17 g/mol Bod tání: 85…89 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007965 CAS číslo: 621-84-1 EINECS: 210-710-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...