Benzyl carbamate 98%
Dodavatel: Thermo Scientific
105890250
105891000
ACRO105890250EA
1210
CZK
ACRO105890250
ACRO105891000
Benzyl carbamate 98%
Benzyl carbamate
Vzorec:
C₈H₉NO₂ MW: 151,17 g/mol Bod tání: 86…89 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007965 CAS číslo: 621-84-1 EINECS: 210-710-4 |
Výsledky specifikačního testu
Appearance | Off-white to light beige Powder or flakes |
Infrared spectrum | Conforms |
Melting point | 86 to 89 °C |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...