Tisk…

(S)-(+)-Methyl-3-hydroxybutyrate ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: Methyl (S)-(+)-3-Hydroxybutyrate, (S)-Methyl 3-hydroxybutanoate, Methyl (S)-(+)-3-hydroxybutyrate

L19996.06 L19996.14
L19996.06EA 3810 CZK
L19996.06 L19996.14
(S)-(+)-Methyl-3-hydroxybutyrate ≥98%
(S)-(+)-Methyl-3-hydroxybutyrate

Purity: 98% CAS No.: 53562-86-0 Molecular Formula: C5H10O3 Formula Weight: 118.13 Flash Point: 73°(163°F) Refractive Index: 1.4220 Boiling Point: 56-58°/11mm Beilstein Reference: 1720583 Optical Rotation: +48.5° (c=1.3 in chloroform) MDL No.: MFCD00064461

Vzorec: C₅H₁₀O₃
MW: 118.13 g/mol
Bod varu: 56…58 °C (11 mmHg)
Hustota: 1.055
Bod vzplanutí: 73 °C (163 °F)
Storage Temperature: Ambient
MDL Number: MFCD00064461
CAS číslo: 53562-86-0
EINECS: 258-628-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR