Tisk…

(R)-(-)-Methyl-3-hydroxybutyrate ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

L14151.03 L14151.06 L14151.14
L14151.03EA 1310 CZK
L14151.03 L14151.06 L14151.14
(R)-(-)-Methyl-3-hydroxybutyrate ≥98%
(R)-(-)-Methyl-3-hydroxybutyrate

Purity: 98% CAS No.: 3976-69-0 Molecular Formula: C5H10O3 Formula Weight: 118.13 Flash Point: 73°(163°F) Refractive Index: 1.4220 Odour: Pleasant Boiling Point: 56-58°/11mm Beilstein Reference: 3648161 Optical Rotation: -48.5° (c=1.3 in chloroform) MDL No.: MFCD00063289

Vzorec: C₅H₁₀O₃
Bod varu: 56…58 °C (11 mmHg)
Hustota: 1,055
Bod vzplanutí: 73 °C (163 °F)
Storage Temperature: Ambient
MDL Number: MFCD00063289
CAS číslo: 3976-69-0
EINECS: 223-610-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR