Ethyl-3-hydroxybutyrate 99%
Dodavatel: Thermo Scientific
Synonyms:
Ethyl 3-hydroxybutanoate, DL-Ethyl-3-hydroxybutyrate, Ethyl 3-hydroxybutyrate
118541000
118545000
118540250
ACRO118541000EA
2730
CZK
ACRO118541000
ACRO118545000
ACRO118540250
Ethyl-3-hydroxybutyrate 99%
Ethyl-3-hydroxybutyrate
Vzorec:
C₆H₁₂O₃ MW: 132,16 g/mol Bod varu: 170 °C (760 mmHg) Hustota: 1,01 g/cm³ Bod vzplanutí: 64 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004545 CAS číslo: 5405-41-4 EINECS: 226-456-2 |
Výsledky specifikačního testu
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4195 to 1.4215 (20°C, 589 nm) |
GC | ≥98.5 % |
Ammonium (NH4) | ≤50 ppm |
Calcium (Ca) | ≤600 ppm |
Iron (Fe) | ≤10 ppm |
Lead (Pb) | ≤50 ppm |
Magnesium (Mg) | ≤100 ppm |
Potassium (K) | ≤200 ppm |
Sodium (Na) | ≤500 ppm |
Oxychloride | passes test |
Insoluble matter | ≤0.005 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...