Tisk…

2,5-Dibromo-3-hexylthiophene, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

456373-5G 456373-25G
SIAL456373-5GEA 9560 CZK
SIAL456373-5G SIAL456373-25G
2,5-Dibromo-3-hexylthiophene, Sigma-Aldrich®
2,5-Dibromo-3-hexylthiophene
Vzorec: C₁₀H₁₄Br₂S
MDL Number: MFCD00274313
CAS číslo: 116971-11-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR