1,2,3,4-Tetrahydroisoquinoline, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
T13005-100G
T13005-25G
SIALT13005-100GEA
4720
CZK
SIALT13005-100G
SIALT13005-25G
1,2,3,4-Tetrahydroisoquinoline, Sigma-Aldrich®
1,2,3,4-Tetrahydroisoquinoline
Vzorec:
C₉H₁₁N MW: 133,19 g/mol |
MDL Number:
MFCD00006896 CAS číslo: 91-21-4 EINECS: 202-050-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...