Diethyl propylphosphonate ≥97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
Diethyl 1-propylphosphonate
A17105.09
A17105.18
A17105.09EA
1350
CZK
A17105.09
A17105.18
Diethyl propylphosphonate ≥97%
Diethyl propylphosphonate
Purity: 97%
CAS No.: 18812-51-6
Molecular Formula: C7H17O3P
Formula Weight: 180.19
Refractive Index: 1.4190
Boiling Point: 94-96°/12mm
MDL No.: MFCD00015139
Vzorec:
C₇H₁₇O₃P MW: 180.19 g/mol Bod varu: 94…96 °C (12 mmHg) Storage Temperature: Ambient |
MDL Number:
MFCD00015139 CAS číslo: 18812-51-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...