Tisk…

5-Fluoro-2-methoxyphenylboronic acid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 5-Fluoro-2-methoxybenzeneboronic acid

M-Clarity™ quality level = MQ100

483540-25G 483540-5G
SIAL483540-25GEA 0 CZK
SIAL483540-25G SIAL483540-5G
5-Fluoro-2-methoxyphenylboronic acid, Sigma-Aldrich®
5-Fluoro-2-methoxyphenylboronic acid
Vzorec: C₇H₈BFO₃
MW: 169,95 g/mol
MDL Number: MFCD01863526
CAS číslo: 179897-94-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR