3-Fluoro-4-methoxyphenylboronic acid ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
(3-Fluoro-4-methoxyphenyl)boronic acid, 3-Fluoro-4-methoxybenzeneboronic acid
L19818.03
L19818.06
L19818.03EA
1960
CZK
L19818.03
L19818.06
3-Fluoro-4-methoxyphenylboronic acid ≥98%
3-Fluoro-4-methoxyphenylboronic acid
Purity: 98+%
CAS No.: 149507-26-6
Molecular Formula: C7H8BFO3
Formula Weight: 169.95
Melting Point: 237-239°
Beilstein Reference: 7370120
Vzorec:
C₇H₈BFO₃ MW: 169,95 g/mol Bod tání: 237…239 °C Storage Temperature: Ambient |
MDL Number:
MFCD00807404 CAS číslo: 149507-26-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...