Tisk…

2-Fluoro-6-methoxyphenylboronic acid ≥98%

Dodavatel: Thermo Scientific
Varování

Synonyms: 2-Fluoro-6-methoxybenzeneboronic acid

H26064.06 H26064.03
H26064.06EA 1760 CZK
H26064.06 H26064.03
2-Fluoro-6-methoxyphenylboronic acid ≥98%
2-Fluoro-6-methoxyphenylboronic acid

Purity: 98% CAS No.: 78495-63-3 Molecular Formula: C7H8BFO3 Formula Weight: 169.95 Melting Point: 124-126° MDL No.: MFCD02179483

Vzorec: C₇H₈BFO₃
MW: 169,95 g/mol
Bod tání: 124…126 °C
Storage Temperature: Ambient
MDL Number: MFCD02179483
CAS číslo: 78495-63-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR