Tisk…

3-(3,4-Dihydroxyphenyl)propionic acid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Dihydrocaffeic acid

M-Clarity™ quality level = MQ100

102601-10G 102601-2.5G 102601-25G
SIAL102601-10GEA 4730 CZK
SIAL102601-10G SIAL102601-2.5G SIAL102601-25G
3-(3,4-Dihydroxyphenyl)propionic acid, Sigma-Aldrich®
3-(3,4-Dihydroxyphenyl)propionic acid
Vzorec: C₉H₁₀O₄
MW: 182,18 g/mol
MDL Number: MFCD00002776
CAS číslo: 1078-61-1
EINECS: 214-083-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR