Tisk…
Isobutylene oxide (2,2-dimethyloxirane, 1,2-epoxy-2-methylpropane), Sigma-Aldrich®
Dodavatel: Merck
Synonyms: Isobutylene oxide
M-Clarity™ quality level = MQ200
531537-10G 531537-50G
SIAL531537-10GEA 5960 CZK
SIAL531537-10G SIAL531537-50G
Isobutylene oxide (2,2-dimethyloxirane, 1,2-epoxy-2-methylpropane), Sigma-Aldrich®
Isobutylene oxide (2,2-dimethyloxirane, 1,2-epoxy-2-methylpropane)
Vzorec: C₄H₈O
MW: 72,11 g/mol
MDL Number: MFCD00066354
CAS číslo: 558-30-5
EINECS: 209-193-8
UN: 2924
ADR: 3,II