Sodium cyanamidate, Sigma-Aldrich®
Dodavatel: Merck
|
Varování
|
Synonyms:
Sodium hydrogencyanamide
M-Clarity™ quality level = MQ100
495905-25G
495905-5G
SIAL495905-25GEA
5430
CZK
SIAL495905-25G
SIAL495905-5G
Sodium cyanamidate, Sigma-Aldrich®
Sodium cyanamidate
Vzorec:
CHN₂Na MW: 64,02 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00066500 CAS číslo: 17292-62-5 EINECS: 241-322-3 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...