Calix[4]arene 98
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
1,3,5,7(1,3)-Tetrabenzenacyclooctaphane-12,32,52,72-tetraol
B21320.03
B21320.06
B21320.03EA
3920
CZK
B21320.03
B21320.06
Calix[4]arene 98
Calix[4]arene
Purity: 98%
CAS No.: 74568-07-3
Molecular Formula: C28H24O4
Formula Weight: 424.50
Melting Point: 315-318°
MDL No.: MFCD00143912
Vzorec:
C₂₈H₂₄O₄ MW: 424.5 Bod tání: 315…318 °C Storage Temperature: Ambient |
MDL Number:
MFCD00233673 CAS číslo: 74568-07-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...