(Iodomethyl)cyclopentane 98% stabilised
Dodavatel: Thermo Fisher Scientific
|
Varování
|
378640010
378640050
378640250
ACRO378640010EA
1800
CZK
ACRO378640010
ACRO378640050
ACRO378640250
(Iodomethyl)cyclopentane 98% stabilised
(Iodomethyl)cyclopentane
Vzorec:
C₆H₁₁I Bod varu: 78 °C (17 mmHg) Hustota: 1,6 g/cm³ Bod vzplanutí: 92 °C Storage Temperature: Ambient |
MDL Number:
MFCD03453190 CAS číslo: 27935-87-1 |
Výsledky specifikačního testu
Appearance | Colorless to yellow to light orange Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5360 to 1.5380 (20°C, 589 nm) |
GC | ≥97.5 % |
Stabilizer | copper powder |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...