Cacodylic acid sodium salt trihydrate, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
M-Clarity™ quality level = MQ200
C4945-100G
C4945-10G
C4945-25G
SIALC4945-100GEA
22430
CZK
SIALC4945-100G
SIALC4945-10G
SIALC4945-25G
Cacodylic acid sodium salt trihydrate, Sigma-Aldrich®
Cacodylic acid sodium salt trihydrate
Vzorec:
(CH₃)₂As(O)ONa·3H₂O Storage Temperature: Ambient |
MDL Number:
MFCD00149079 CAS číslo: 6131-99-3 EINECS: 204-708-2 UN: 1688 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...