Cacodylic acid sodium salt trihydrate ≥98%, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
M-Clarity™ quality level = MQ200
C0250-100G
C0250-10G
C0250-25G
C0250-500G
SIALC0250-100GEA
9570
CZK
SIALC0250-100G
SIALC0250-10G
SIALC0250-25G
SIALC0250-500G
Cacodylic acid sodium salt trihydrate ≥98%, Sigma-Aldrich®
Cacodylic acid sodium salt trihydrate
Vzorec:
(CH₃)₂As(O)ONa·3H₂O MW: 213,94 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00149079 CAS číslo: 6131-99-3 EINECS: 204-708-2 UN: 1688 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...