Cacodylic acid sodium salt trihydrate 98%, pure
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
Dimethylarsinic acid sodium salt trihydrate, Sodium cacodylate trihydrate, Dimethylarsonic acid sodium salt trihydrate, Sodium dimethylarsinate trihydrate
214970250
214971000
214975000
214970100
ACRO214970250EA
2820
CZK
ACRO214970250
ACRO214971000
ACRO214975000
ACRO214970100
Cacodylic acid sodium salt trihydrate 98%, pure
Cacodylic acid sodium salt trihydrate
Vzorec:
(CH₃)₂As(O)ONa·3H₂O MW: 213.94 Bod tání: 77…80 °C Storage Temperature: Ambient |
MDL Number:
MFCD00149079 CAS číslo: 6131-99-3 EINECS: 204-708-2 UN: 1688 ADR: 6.1 II |
Výsledky specifikačního testu
Appearance | White Crystalline powder or crystals |
Infrared spectrum | Conforms |
Titration with HCl | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...