Tisk…

(R)-(+)-4-Benzyl-2-oxazolidinone, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: (R)-4-Benzyl-2-oxazolidinone

M-Clarity™ quality level = MQ100

300977-5G 300977-25G
SIAL300977-5GEA 6050 CZK
SIAL300977-5G SIAL300977-25G
(R)-(+)-4-Benzyl-2-oxazolidinone, Sigma-Aldrich®
(R)-(+)-4-Benzyl-2-oxazolidinone
Vzorec: C₁₀H₁₁NO₂
MW: 177,2 g/mol
MDL Number: MFCD00010846
CAS číslo: 102029-44-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR