(S)-(-)-4-Isopropyl-2-oxazolidinone, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
(4S)-(-)-Isopropyl-2-oxazolidinone, (4S)-(-)-4-Isopropyl-2-oxazolidinone, (S)-4-Isopropyl-2-oxazolidinone, (4S)-2-Oxo-4-(prop-2-yl)-1,3-oxazolidine, (4S)-4-(Prop-2-yl)-1,3-oxazolidin-2-one
M-Clarity™ quality level = MQ100
298883-1G
298883-5G
SIAL298883-1GEA
2390
CZK
SIAL298883-1G
SIAL298883-5G
(S)-(-)-4-Isopropyl-2-oxazolidinone, Sigma-Aldrich®
(S)-(-)-4-Isopropyl-2-oxazolidinone
Vzorec:
C₆H₁₁NO₂ MW: 129.16 |
MDL Number:
MFCD00010847 CAS číslo: 17016-83-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...